Learn More
MSC Koch Filter DuraMAX 4v Air Filter, Microfiberglass, MERV 11, 1 ea
Catalog No. 5000507750
Encompass
The DuraMAX 4v is a high-capacity, rigid, extended surface air filter engineered for maximum performance and prolonged service life in demanding environments. Featuring a V-bank mini-pleat design, this filter is optimized for filtration systems requiring high velocities or variable air volume (VAV) applications. The filter utilizes a lightweight, extremely rigid plastic frame that allows for easy incineration after use.
  • High capacity mini-pleat design reduces resistance to airflow and energy costs
  • Constructed with high-efficiency microfiberglass media
  • Superior dust-holding capacity for extended filter life
  • Compliant with ASHRAE 52.2-2012 and UL 900 standards
  • Incinerable all-plastic construction
  • Temperature rated to 200°F
Catalog No. 50-005-07750 Supplier MSC Supplier No. 85094365
The DuraMAX 4v is a high-capacity, rigid, extended surface air filter engineered for maximum performance and prolonged service life in demanding environments. Featuring a V-bank mini-pleat design, this filter is optimized for filtration systems requiring high velocities or variable air volume (VAV) applications. The filter utilizes a lightweight, extremely rigid plastic frame that allows for easy incineration after use.
  • High capacity mini-pleat design reduces resistance to airflow and energy costs
  • Constructed with high-efficiency microfiberglass media
  • Superior dust-holding capacity for extended filter life
  • Compliant with ASHRAE 52.2-2012 and UL 900 standards
  • Incinerable all-plastic construction
  • Temperature rated to 200°F