Learn More
Glass Expansion DuraMist DC Nebulizer 0.4mL/min, Includes NFT-050 UniFit Connector with 0.50mm ID x 1.3mm OD x 700mm long tubing (1 piece) and 70-803-0969 Gas Fitting with Ratchet Nebulizer connector
SDP
Catalog No. 501907370
Encompass_Preferred
DuraMist DC Nebulizer 0.4mL/min, includes UniFit Connector with 0.50mm ID x 1.3mm OD x 700mm long tubing (1 pc) and DC Gas Fitting Type 11 with ratchet connector (1 pc). Compatible with Analytik Jena ICP-MS and Leeman.Optimum nebulizer gas flow rate is 1.0L/min & sample flow rate is 0.4mL/min. Recommended not to exceed optimum 0.4mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 1.0L/min gas flow rate. DuraMist is an economical inert nebulizer for high-precision analysis & the highest tolerance to TDS, up to 30%. Made from PEEK, it tolerates up to 5% HF.Replacement parts include P/Ns NC1625341 and 502703076.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N 501907353). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7370 Supplier Glass Expansion Supplier No. A111DM04
DuraMist DC Nebulizer 0.4mL/min, includes UniFit Connector with 0.50mm ID x 1.3mm OD x 700mm long tubing (1 pc) and DC Gas Fitting Type 11 with ratchet connector (1 pc). Compatible with Analytik Jena ICP-MS and Leeman.Optimum nebulizer gas flow rate is 1.0L/min & sample flow rate is 0.4mL/min. Recommended not to exceed optimum 0.4mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 1.0L/min gas flow rate. DuraMist is an economical inert nebulizer for high-precision analysis & the highest tolerance to TDS, up to 30%. Made from PEEK, it tolerates up to 5% HF.Replacement parts include P/Ns NC1625341 and 502703076.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N 501907353). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001