Learn More
Glass Expansion DuraMist DC Nebulizer 1mL/min with 0.25mm ID x 1.6mm OD sample tube
SDP
Catalog No. 5000556493
Encompass_Preferred
DuraMist DC Nebulizer 1mL/min with 0.25mm ID x 1.6mm OD sample tube Sourced from Glass Expansion
Catalog No. 50-005-56493 Supplier Glass Expansion Supplier No. A1107DM1Q
DuraMist DC Nebulizer 1mL/min with 0.25mm ID x 1.6mm OD sample tube Sourced from Glass Expansion