Learn More
Glass Expansion DuraMist DC Nebulizer 1mL/min compatible with Thermo Pro, 6000 series and 7000 series ICP-OES.
SDP
Catalog No. 502703015
Encompass_Preferred
DuraMist DC Nebulizer 1.0mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 pc) and DC Gas Fitting Type 31 with ratchet connector (1 pc). Compatible with Thermo Pro, 6000 Series and 7000 Series ICP-OES.Optimum nebulizer gas flow rate is 0.7L/min & sample flow rate is 1.0mL/min. Recommended not to exceed optimum 1.0mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 0.7L/min gas flow rate. DuraMist is an economical inert nebulizer for high-precision analysis & the highest tolerance to TDS, up to 30%. Made from PEEK, it tolerates up to 5% HF.Replacement parts include P/Ns NC1625341 and 501907300.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3015 Supplier Glass Expansion Supplier No. A3107DM1
DuraMist DC Nebulizer 1.0mL/min, includes UniFit Connector with 0.50mm ID x 1.6mm OD x 700mm long tubing (1 pc) and DC Gas Fitting Type 31 with ratchet connector (1 pc). Compatible with Thermo Pro, 6000 Series and 7000 Series ICP-OES.Optimum nebulizer gas flow rate is 0.7L/min & sample flow rate is 1.0mL/min. Recommended not to exceed optimum 1.0mL/min sample flow rate; however, lower gas & sample flow rates are OK. The backpressure should read ~40 psi at 0.7L/min gas flow rate. DuraMist is an economical inert nebulizer for high-precision analysis & the highest tolerance to TDS, up to 30%. Made from PEEK, it tolerates up to 5% HF.Replacement parts include P/Ns NC1625341 and 501907300.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) & Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001