Learn More
Perkin Elmer US LLC DuraMist Nebulizer, 0.4 mL/min for ELAN/NexION 300/350
Catalog No. 502104658
Encompass_Preferred
This DuraMist Nebulizer has a sample uptake of 0.4 mL/min at 1 L/min of Argon flow.
Catalog No. 50-210-4658 Supplier Perkin Elmer US LLC Supplier No. N8121046
Add to Cart
Add to Cart
Request a Quote
This DuraMist Nebulizer has a sample uptake of 0.4 mL/min at 1 L/min of Argon flow.