Learn More
Perkin Elmer US LLC DuraMist Nebulizer, 0.4 mL/min for ICP-OES
Catalog No. 502104218
Encompass_Preferred
Air Flow Rate: 0.7 L/min The DuraMist is made from PEEK and can tolerate a wide variety of solvents including HF* High sensitivity and excellent precision due to concentric design and rigid construction High tolerance to dissolved solids (typically up to 30%) due to smooth surface of capillary and nebulizer tip
Catalog No. 50-210-4218 Supplier Perkin Elmer US LLC Supplier No. N0777713
Air Flow Rate: 0.7 L/min The DuraMist is made from PEEK and can tolerate a wide variety of solvents including HF* High sensitivity and excellent precision due to concentric design and rigid construction High tolerance to dissolved solids (typically up to 30%) due to smooth surface of capillary and nebulizer tip