Learn More
Perkin Elmer US LLC DuraMist Nebulizer 1.0 mL/min for ICP-MS
Catalog No. 502104425
Encompass_Preferred
The DuraMist nebulizer is recommended for samples containing up to 5% HF. For samples with higher concentrations of HF, the OpalMist PFA nebulizer is recommended. This DuraMist Nebulizer has a sample uptake of 1.0 mL/min at 1 L/min of Argon flow.
Catalog No. 50-210-4425 Supplier Perkin Elmer US LLC Supplier No. N8151078
The DuraMist nebulizer is recommended for samples containing up to 5% HF. For samples with higher concentrations of HF, the OpalMist PFA nebulizer is recommended. This DuraMist Nebulizer has a sample uptake of 1.0 mL/min at 1 L/min of Argon flow.