Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, GL 45 Thread Neck, GL 32 Thread Side Arm
Shop All DWK Life Sciences Products

Description
Aspirator laboratory bottle with screw closure, complete with PTFE key stopcock and GL 45 red PBT cap.
- Supplied ready to use
- DURAN™ glass and PTFE key stopcock outlet
- Made of virtually inert, chemically resistant DURAN™ glass
- Fully compatible with all the GL 45 threads and therefore highly versatile
- Tabulator with GL 32 thread
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifications
Specifications
| Capacity (Metric) | 2000 mL |
| For Use With (Application) | Storage and dispensing of liquids and reagents |
| Material | Borosilicate Glass (3.3), Silicone, PTFE |
| No. per Pack | 1/Pk. |
| Neck Finish | Thread Size GL 45 |
| Diameter (Metric) | 136 mm |
| Autoclavable | Autoclavable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.