Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, GL 45 Thread Neck, GL 32 Thread Side Arm
Shop All DWK Life Sciences Products

Description
- Supplied ready to use with PTFE key stopcock and GL 45 red PBT screw cap
- Made of virtually inert, chemically resistant DURAN™ borosilicate glass
- Neck with DIN thread GL 45; tabulator with GL 32 thread
- Fully compatible with all GL 45 thread accessories for high versatility
- Easy-to-read scale with graduation marks to estimate content
- Fired-on, highly durable white ceramic scale markings
- Dosing of liquids possible via the PTFE spindle stopcock
Specifications
Specifications
| Capacity (Metric) | 10,000 mL |
| Color | Clear |
| For Use With (Application) | Storage and dispensing of liquids and reagents |
| Material | Borosilicate Glass (3.3), Silicone, PTFE |
| No. per Pack | 1/Pk. |
| Sterility | Non-sterile |
| Neck Finish | Thread Size GL 45 |
| Diameter (Metric) | 230 mm |
| Autoclavable | Autoclavable |
| Product Type | Aspirator Bottle |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.