Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC DURAN BAFFLED WM BOTTLE GLS80
Supplier:  ATLANTIC BIOPHARM SOLUTIONS LLC ABS1154R36
Encompass_Preferred
DURAN baffled wide mouth Bottle GLS 80, 1000 ml, clear glass, supplied complete with GLS 80 polypropylene pouring ring and GLS 80 cap
Catalog No. NC2871933