Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Duran™ EPDM O‐Ring Gasket for GLS 80™ Bottles and Stirred Reactors
SDP

Catalog No. 12141615 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12-141-615 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-141-615 Supplier DWK Life Sciences Supplier No. 1152913
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EPDM O‐ring gasket provides a reliable seal between GLS 80™ stirred reactors and glass bottles, helping prevent leaks, maintain system integrity, and support safe, efficient laboratory processing.

  • Manufactured from EPDM
  • Improved sealing with GLS 80™ bottles and connector caps
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Outside 78 mm
For Use With (Application) Liquid‐tight Sealing
For Use With (Equipment) For GLS 80 stirred reactors and laboratory bottles
Type Gasket
Color Black
Quantity 5
Diameter (English) Outside 3.07 in.
Material EPDM
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.