Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ DURAN™ GL 45 Laboratory Glass Bottles With PP Screw Cap and Pouring Ring
GSA_VASDP

Catalog No. 09841180 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
2000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09-841-180 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-841-180 Supplier DWK LIFE SCIENCES™ Supplier No. 218016357
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DURAN™ GL 45 Laboratory Glass Bottles With PP Screw Cap and Pouring Ring combine proven DURAN™ borosilicate glass quality with a blue PP screw cap (GL 45, integral lip seal) and PP pouring ring for drip-free, safe pouring. Ideal for storage, sample preparation and transport.

  • DURAN™ borosilicate glass bottle with GL 45 thread
  • Supplied complete with blue polypropylene (PP1) screw cap featuring integral lip seal
  • Polypropylene (PP1) pouring ring enables drip-free pouring and clean, safe working
  • Service temperature of screw cap and pouring ring: up to +140°C
  • Typical applications: storage, sample preparation and transport
  • Select sizes available with retrace code

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2000 mL
Color Clear
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Material Borosilicate Glass (Type I)
Sterility Non-sterile
Thread Size (Metric) GL 45
Closure Included Yes
Graduated Yes
Mouth Threaded
Height (Metric) 265 mm
Closure Size GL 45
Diameter (Metric) 136 mm
Autoclavable Autoclavable
Capacity (English) 68 oz.
Shape Square
Disposable Reusable
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.