Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ GL 45 Thread Adaptor for Bottle Top Vacuum Filtration Unit
SDP

Catalog No. 14209157 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-209-157 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-209-157 Supplier DWK Life Sciences Supplier No. 291195601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DURAN™ GL 45 Thread Adaptor for Bottle Top Vacuum Filtration Unit. The DURAN™ GL 45 Thread Adaptor for Bottle Top Vacuum Filtration Unit connects GL 45 external to GL 56 internal threads. Made from PTFE, this white adaptor is an essential accessory for DURAN™ bottle top vacuum filtration setups

  • Accessory for DURAN™ bottle top vacuum filtration unit
  • GL 45 external thread connects to GL 56 internal thread
  • Material: PTFE
  • Colour: White
  • Pack quantity: 1
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material PTFE
Sterility Non-sterile
For Use With (Equipment) Bottle top vacuum filtration unit (GL 45 external, GL 56 internal)
Autoclavable Autoclavable
Disposable Reusable
Product Type Thread Adapter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.