Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ GLS 80 Screw Cap for Laboratory Glass Bottles
SDP

Catalog No. 09841273 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
1 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
09-841-273 1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-841-273 Supplier DWK Life Sciences Supplier No. 1112716
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The DURAN™ GLS 80 Screw Cap for Laboratory Glass Bottles allows quick, secure opening and closing of DURAN™ GLS 80 wide-mouth bottles with only a three-quarter turn. Made from polypropylene with a lip seal for a reliable, tight closure.

  • Designed for use with DURAN GLS 80 wide-mouth laboratory bottles
  • Opens and closes with only a three-quarter turn for fast, convenient access
  • Made from polypropylene (PP) for chemical resistance
  • Blue colour for easy identification
  • Fitted with a lip seal to ensure a tight, secure closure
  • GLS 80 thread size
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polypropylene
Sterility Non-sterile
Thread Size (Metric) 1 mm
Neck Finish 1 mm
For Use With (Equipment) Laboratory glass bottles
Autoclavable Autoclavable
Disposable Reusable
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.