Learn More
DWK Life Sciences DURAN™ GLS 80 Screw Cap with PTFE-Coated Silicone Seal for Laboratory Glass Bottles
Shop All DWK Life Sciences Products

Description
- GLS 80 screw cap designed for use with DURAN laboratory glass bottles
- Made from white polysulfone (PSU) for chemical and thermal resistance
- PTFE-coated silicone seal for a secure, reliable closure
- Compatible with GLS 80 thread size
Specifications
Specifications
| Color | White |
| Material | PSU |
| Sterility | Non-sterile |
| Thread Size (Metric) | 1 mm |
| Neck Finish | 1 mm |
| For Use With (Equipment) | Laboratory glass bottles |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Product Type | Screw Cap with Silicone Seal |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.