Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ GLS 80™ Laboratory bottle, Double Walled (Jacketed), Wide Mouth, Clear, Graduated
SDP

Catalog No. 02900161 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-900-161 500 mL
02-900-162 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-900-161 Supplier DWK Life Sciences Supplier No. 242564451
Add to Cart
Add to Cart

The double walls allows the temperature of the contents of the wide mouth GLS 80™ bottle to be controlled.

The DURAN™ double walled, wide mouth bottles GLS 80™ incorporate an integral jacket that isolates the contents from the external environment. Heated or cooled liquids can be circulated through the jacket to control the temperature within the screw topped DURAN™ bottle. The DURAN™ double walled bottles offer a sealable and more flexible alternative to open topped jacketed beakers.

  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • Ensured consistent heating or cooling due to the positions of the 10 mm PTFE inlet/outlet hose connectors around the jacket
  • Easy-to-read graduated volume scale on the inner vessel
  • The GL 18 outlet hose connectors are removable for increased safety
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
For Use With (Application) Simple, low cost, jacketed chemostat or bioreactor for temperature-controlled reactions such as enzyme digestions; Storage at a constant temperature; Working with endo- or exo-thermic reagents; Large cold trap or condenser when used with dry ice in the inner container
Includes With screw cap and pouring ring from PP (blue)
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Thread Size (Metric) GLS 80
Closure Material Polypropylene
Diameter (Metric) 110 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.