Learn More
DWK LIFE SCIENCES™ DURAN™ GLS 80™ Laboratory Bottle, Double Walled (Jacketed), Wide Mouth, Clear, Graduated
Shop All DWK Life Sciences Products

Description
- Integral jacket isolates bottle contents from the external environment for precise temperature control
- Heated or cooled liquids can be circulated through the jacket to control the temperature within the screw-topped bottle
- Wide GLS 80 mouth provides easy access to bottle contents
- 10 mm PTFE inlet/outlet hose connectors positioned around the jacket ensure consistent heating or cooling
- Easy-to-read graduated volume scale on the inner vessel
- Removable GL 18 outlet hose connectors for increased safety
- Sealable and more flexible alternative to open-topped jacketed beakers
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Color | Clear |
| For Use With (Application) | Simple, low cost, jacketed chemostat or bioreactor for temperature-controlled reactions such as enzyme digestions; Storage at a constant temperature; Working with endo- or exo-thermic reagents; Large cold trap or condenser when used with dry ice in the inner container |
| Includes | With screw cap and pouring ring from PP (blue) |
| Material | Borosilicate Glass (3.3) |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
| Thread Size (Metric) | GLS 80 |
| Closure Included | Yes |
| Graduated | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.