Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ DURAN™ GLS 80™ Laboratory Bottle Only, Wide Mouth, Amber, GLS 80™ Thread, USP/EP (3.2.1) Light Transmission, Graduated
SDP

Catalog No. 09841218 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
20,000 mL
250 mL
3500 mL
500 mL
5000 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09-841-218 3500 mL
09-841-213 500 mL
09-841-214 1000 mL
02-900-150 10,000 mL
02-900-151 20,000 mL
09-841-216 5000 mL
09-841-217 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 09-841-218 Supplier DWK LIFE SCIENCES™ Supplier No. 218666901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DURAN™ GLS 80™ Laboratory Bottle Only, Wide Mouth, Amber, GLS 80™ Thread, USP/EP (3.2.1) Light Transmission, Graduated. DURAN™ GLS 80™ Laboratory Bottle Only, Wide Mouth, Amber offers UV protection to approx. 500 nm, USP/EP Type I borosilicate glass 3.3, wide-mouth GLS 80™ thread, graduated scale, a

  • Moulded from high-purity USP/EP Type I borosilicate glass 3.3 with excellent thermal performance and high chemical resistance
  • Fused external amber layer protects light-sensitive contents from photodegradation; UV protection to approx. 500 nm meets USP/EP standards and lasts the lifetime of the bottle
  • Internal surfaces retain standard DURAN™ glass properties as colour is applied externally only
  • Wide-mouth GLS 80TM thread enables rapid opening and closing in less than one turn and easy access to bottle contents
  • Optimum filling, pouring, and particularly easy cleaning; large base area provides stability
  • Spoons, spatulas, tweezers, and even large-volume funnels can be inserted with ease
  • Easy-to-read graduated volume scale and marking field in fired-on, highly durable white ceramic enamel
  • Permanent Retrace Code on each bottle for manufacturing lot traceability
  • Supplied complete with blue polypropylene cap and drip-free pouring ring; all components fully autoclavable at 121°C or 134°C
  • Robust ISO 4796-1 design provides a long working life
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 3500 mL
Color Amber
For Use With (Application) Autoclaving of of light sensitive media and reagents; Preparation of light sensitive media and reagents; Sample collection of light sensitive materials; Culturing vessel; Storage of light sensitive media and reagents; Transport and shipping of light sensitive media and reagents; Liquid handling reservoir
Includes Without screw cap and pouring ring from PP (blue)
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Sterility Non-sterile
Thread Size (Metric) GLS 80
Closure Included Yes
Graduated Yes
Mouth Wide
Closure Material Polypropylene
Closure Size GLS 80
Diameter (Metric) 160 mm
Autoclavable Autoclavable
Capacity (English) 118 oz.
Shape Round
Disposable Reusable
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.