Learn More
DWK LIFE SCIENCES™ DURAN™ GLS 80™ Laboratory Bottle with Screw Cap and Pouring Ring, Wide Mouth, Amber, GLS 80™ Thread
Shop All DWK Life Sciences Products

Description
- Made from high-purity USP/EP Type I borosilicate glass 3.3 with excellent thermal performance and high chemical resistance
- Fused external amber layer provides UV protection up to approximately 500 nm, meeting USP/EP standards for the lifetime of the bottle
- Internal surfaces retain standard DURAN™ glass properties as colour is applied externally only
- Wide GLS 80TM mouth enables easy access to bottle contents and insertion of large-volume funnels
- Special GLS 80TM thread design allows rapid opening and closing in less than one full turn
- Optimised geometry supports easy filling, pouring, and cleaning; spoons, spatulas, and tweezers can be used with ease
- Large base area provides stability during use
- Easy-to-read graduated volume scale and marking field in fired-on, highly durable white ceramic enamel
- Permanent Retrace Code on each bottle for manufacturing lot traceability
- Supplied complete with blue polypropylene screw cap and drip-free pouring ring rated for use up to 140°C
- Fully autoclavable at 121°C or 134°C; conforms to ISO 4796-1
Specifications
Specifications
| Capacity (Metric) | 3500 mL |
| Color | Amber |
| For Use With (Application) | Autoclaving of of light sensitive media and reagents; Preparation of light sensitive media and reagents; Sample collection of light sensitive materials; Culturing vessel; Storage of light sensitive media and reagents; Transport and shipping of light sensitive media and reagents; Liquid handling reservoir |
| Includes | Blue Polypropylene Screw Cap and Pouring Ring |
| Material | Borosilicate Glass (3.3) |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
| Thread Size (Metric) | GLS 80 |
| Closure Included | Yes |
| Graduated | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.