Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, GLS 80™ Thread, Graduated, with Screw Cap and Pouring Ring
SDP

Catalog No. 02900153 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
30 L
50,000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-900-153 30 L
02-900-152 50,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-900-153 Supplier DWK LIFE SCIENCES™ Supplier No. 1200265
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, GLS 80™ Thread, Graduated, with Screw Cap and Pouring Ring is made from USP/EP Type I borosilicate glass 3.3 with a wide GLS 80 neck for easy access, fast opening, and autoclavable use up to 134°C.

Wide mouth, multifunctional glass laboratory bottles.

  • Moulded from high-purity, clear USP/EP Type I borosilicate glass 3.3 for excellent thermal performance and high chemical resistance
  • Wide GLS 80 mouth provides easy access to bottle contents and accommodates large-volume funnels, spoons, spatulas and tweezers
  • Special GLS 80 thread design enables rapid opening and closing in less than one full turn
  • Optimum filling and pouring geometry with particularly easy cleaning due to wide neck diameter
  • Robust ISO 4796-1 design for a long working life
  • Easy-to-read graduated volume scale and marking field in fired-on, highly durable white ceramic enamel
  • Permanent Retrace Code on each bottle for manufacturing lot traceability
  • Large base area provides excellent stability
  • Supplied complete with blue polypropylene screw cap (integral lip seal) and drip-free polypropylene pouring ring for clean, safe working up to 140°C
  • Fully autoclavable at 121°C or 134°C
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 30 L
Color Clear
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Includes Blue Polypropylene Screw Cap and Pouring Ring
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Sterility Non-sterile
Thread Size (Metric) GLS 80
Closure Color Blue
Closure Included Yes
Graduated Yes
Mouth Wide
Fit for Purpose Certified Quality
Height (Metric) 548 mm
Closure Liner Linerless
Closure Material Polypropylene
Closure Size GLS 80
Diameter (Metric) 340 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 7.93 gal.
Shape Round
Disposable Reusable
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.