Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, Protect Coated, GLS 80™ Thread, Graduated
SDP

Catalog No. 09841221 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
3500 mL
5000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09-841-221 3500 mL
09-841-203 1000 mL
09-841-205 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-841-221 Supplier DWK LIFE SCIENCES™ Supplier No. 218656958

missing translation for 'validMainframeMsg'

DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, Protect Coated, GLS 80™ Thread, Graduated combines USP/EP Type I borosilicate glass 3.3 with a clear PU safety coating for scratch, splinter and UV protection. Autoclavable and ISO 4796-1 compliant.

  • USP/EP Type I borosilicate glass 3.3 interior combined with a clear PU plastic protective safety coating on the exterior
  • Protect safety coating meets ASTM E1522 requirements; provides scratch, leak and splinter protection (applies to bottles 5000 mL and under)
  • UV protection up to approximately 380 nm due to external coating
  • Wide GLS 80 mouth provides easy access to bottle contents and allows insertion of large-volume funnels, spoons, spatulas and tweezers
  • Special GLS 80 thread design enables rapid opening and closing in less than one full turn
  • Optimum filling, pouring and particularly easy cleaning
  • Stability due to large base area
  • Easy-to-read graduated volume scale and marking field in permanent white enamel
  • Permanent Retrace Code on each bottle for manufacturing lot traceability
  • Fully autoclavable at 121°C or 134°C
  • Robust ISO 4796-1 design for long working life
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 3500 mL
Color Clear
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Includes With screw cap and pouring ring from PP (blue)
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Sterility Non-sterile
Thread Size (Metric) 80
Closure Included Yes
Graduated Yes
Mouth Wide
Closure Material Polypropylene
Closure Size GLS 80
Diameter (Metric) 160 mm
Autoclavable Autoclavable
Capacity (English) 118 oz.
Shape Round
Disposable Reusable
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.