Learn More
DWK LIFE SCIENCES™ DURAN™ GLS 80™ Production and Storage Bottle, Carboys, Clear, GLS 80™ Thread, Graduated
Shop All DWK Life Sciences Products

Description
- Thick-walled borosilicate 3.3 glass construction for superior mechanical resistance and durability
- Manufactured from Type I glass conforming to USP, EP, and JP pharmacopoeia standards - ideal for pharmaceutical production
- Manufactured from inorganic materials; certified BSE/TSE free
- Retrace Code included for batch traceability and conformance certification
- Suitable for high-temperature sterilization, depyrogenation, and autoclaving
- Flat, robust base supports use of large magnetic stir bars for mixing processes
- Large, permanent, easy-to-read white ceramic enamel graduation marks for accurate volume estimation
- Wide GLS 80TM neck diameter enables easy filling, pouring, and cleaning
- Supplied without screw cap or pouring ring; compatible with GLS 80TM caps and connector systems
- Available with customized logos, identification labeling, or graduations
Specifications
Specifications
| Capacity (Metric) | 10 L |
| Color | Clear |
| For Use With (Application) | Pharmacutical manufacturing; Autoclaving of media and reagents; Autoclaving of media and reagents; Preparation of media and reagents |
| Material | Borosilicate Glass (3.3) |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
| Thread Size (Metric) | GLS 80 |
| Closure Included | No |
| Graduated | No |
| Mouth | Wide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.