Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ GLS 80™ Screw Cap with Four Ports
SDP

Catalog No. 09841314 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GLS 80
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
09-841-314 GLS 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-841-314 Supplier DWK Life Sciences Supplier No. 293109103
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The DURAN™ GLS 80™ Screw Cap with Four Ports provides a flexible, modular system for safe liquid and gas transfer within closed, sterile environments. Compatible with five tubing diameters (3.2 to 12.0 mm) and fully compatible with all GLS 80 threads.

  • Flexible modular GLS 80 connection system for safe transfer of liquid or gas media within a closed and sterile environment
  • Four ports with GL 18 thread accept five interchangeable insert diameters: 3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm
  • Tubes are fed through ports and secured with inserts and screw connections
  • Sterile pressure equalisation is possible by using the syringe membrane filter (pressure equalisation kit available separately)
  • Unused ports can be sealed with a closed-top GL 18 screw cap (blanking cap)
  • Fully compatible with all GLS 80 threads, offering high versatility across the DURAN™ bottle range
  • Constructed from PP and PTFE; reduces evaporation during media transfer
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Description DURAN GLS 80 screw cap with four ports (GL 18 thread) Blue and grey Polypropylene with white EPDM O-ring
For Use With (Application) Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments; Photo-bioreactor
Material Polypropylene, PTFE, EPDM
Quantity 2/Pk.
Sterility Non-sterile
Thread Size (Metric) GLS 80
Neck Finish 93 mm
Diameter (Metric) 87 mm
Autoclavable Autoclavable
Disposable Reusable
Product Type Bottle Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.