Learn More
DWK Life Sciences DURAN™ Inserts for Screw Cap GL 18
SDP
Made from PP and PTFE this flexible modular system - five different hose diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected.
Supplier:  DWK Life Sciences 1160173
Catalog No. 09-841-147
Add to Cart
Description
Description
  • Typical applications: safe transfer of liquid media within a closed and sterile system
  • Sterile pressure equalization is possible by using the membrane filter
  • Unused ports can be provided with a blind cap (evaporation is reduced).
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions