Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ Laboratory Premium Caps
SDP

Catalog No. 09841064 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
09-841-064 GL 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-841-064 Supplier DWK Life Sciences Supplier No. 1129600
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DURAN™ Laboratory Premium Caps are molded from dye-free polymer to minimise leaching risk, featuring a colorless PTFE-coated silicone seal for high leak tightness and a wide usable temperature range of -196°C to +260°C.

  • Wide usable temperature range from -196°C to +260°C
  • High chemical resistance suitable for aggressive materials
  • Dye-free polymer construction minimises risk of leaching
  • Colorless PTFE-coated silicone sealing disk ensures high leak tightness
  • Ideal for pharmaceutical industry applications, storage of aggressive materials, and demanding sterilisation processes including hot air sterilisation and depyrogenation
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material PFA
Sterility Non-sterile
Thread Size (Metric) GL 25
Neck Finish 12 mm
Height (Metric) 32 mm
For Use With (Equipment) Premium bottle
Diameter (Metric) 22 mm
Autoclavable Autoclavable
Disposable Reusable
Product Type Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.