Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ Mulitport Connector Caps GL 25
SDP

Catalog No. 14209163 Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
Connector Cap (2-port)
Connector Cap (3-port)
Connector Cap (4-port)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
14-209-163 Connector Cap (2-port)
14-209-164 Connector Cap (3-port)
14-209-165 Connector Cap (4-port)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-209-163 Supplier DWK Life Sciences Supplier No. 292601301
Add to Cart
Add to Cart

2, 3 or 4 port connector caps for GL 25 DURAN™ bottles

  • GL 25 thread fits 10 or 25mL DURAN bottles
  • Barbed male hose connector suitable for tubing with a 3.2mm (1/8 in.) ID (such as PharMed™ BPT Tubing™)
  • Available with 2, 3, or 4 surgical grade stainless steel (316L) barbed tubing ports
  • Certified construction materials as approved for food contact
  • High temperature resistance (up to 180°C)
  • Autoclavable at 121°C / 15 minutes)
  • Reusable with a long service life
  • With red PBT screw cap and PTFE insert
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Low volume sampling, Media supply for small bioreactors, Fed-batch bioreactor, Perfusion circuit for cell culture, Oxygenation / aeration of very small samples, Use in a chemistry micro-reactor
Includes PTFE insert, 2 stainless steel barbs
Material PBT
Thread Size (Metric) GL 25
Neck Finish 9 mm
Product Type Connector Cap (2-port)
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.