Learn More
DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT
Shop All DWK Life Sciences Products

Description
- Open topped cap with central aperture compatible with bromobutyl stoppers, silicone piercing septa and glassware connections
- Red PBT material offers excellent temperature and chemical resistance, superior to standard PP caps
- Glass-reinforced PBT provides high mechanical strength
- Compatible with standard GL bottle threads
- Autoclavable for repeated sterilisation use
Specifications
Specifications
| Color | Red |
| Diameter (Metric) Inner | Bore Size 20 mm |
| For Use With (Application) | Aseptic reagent delivery and culture sampling using a syringe and needle |
| Material | PBT |
| Quantity | 10/Pk. |
| Screw Material | Polybutylene Terephthalate (PBT) with glass fiber |
| Sterility | Non-sterile |
| Thread Size (Metric) | GL 32 |
| Closure Material | PBT with Glass Fiber |
| Diameter (Metric) | 42 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.