Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ DURAN™ Original GL 45 Laboratory Bottle, with High Temperature Screw Cap, Clear, Graduated, DIN 168-1 Thread
SDP

Catalog No. 09841303 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
500 mL
5000 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09-841-303 1000 mL
09-841-300 100 mL
09-841-301 250 mL
09-841-302 500 mL
09-841-304 2000 mL
09-841-305 5000 mL
09-841-306 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 09-841-303 Supplier DWK LIFE SCIENCES™ Supplier No. 218015414
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DURAN™ Original GL 45 Laboratory Bottle, with High Temperature Screw Cap, Clear, Graduated, DIN 168-1 Thread is moulded from USP/EP Type I borosilicate glass 3.3 and supplied with a red PBT GL 45 cap, PTFE-faced silicone liner, and ETFE pouring ring. Fully autoclavable at 121 or 134°C.

  • Moulded from high-purity, clear USP/EP Type I DURAN™ borosilicate glass 3.3 for excellent thermal performance and high chemical resistance.
  • Conforms to ISO 4796-1 design for a robust, long working life.
  • Supplied complete with red polybutylene terephthalate (PBT) GL 45 threaded screw cap with PTFE-faced silicone liner and ETFE pouring ring for drip-free pouring.
  • High temperature PBT cap and ETFE pouring ring offer greater thermal and chemical resistance than equivalent polypropylene components.
  • Easy-to-read graduated volume scale and marking field in permanent white enamel.
  • Permanent Retrace Code on each bottle for full manufacturing lot traceability.
  • All components fully autoclavable at 121 or 134°C and suitable for dry heat sterilization up to 180°C.
  • All components fully compliant with current international food contact regulations including (EU) No 10/2011.
  • Suitable for long-term storage of media, chemicals, and solvents.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Color Clear
For Use With (Application) Suitable for dry heat sterilization; Autoclaving of liquid media; Storage of corrosive reagents; Sampling; Storage of media and reagents; Transport and shipping
Includes With high temperature resistant PBT screw cap and ETFE pouring ring
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 45
Closure Included Yes
Graduated Yes
Mouth Threaded
Closure Material PBT with ETFE Pouring Ring
Closure Size GL 45
Diameter (Metric) 101 mm
Autoclavable Autoclavable
Capacity (English) 34 oz.
Shape Round
Disposable Reusable
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.