Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ Original GL 45 Laboratory Bottle, with High Temperature Screw cap, Clear, with DIN 168-1 Thread, Clear, Graduated
SDP

Catalog No. 09841305 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
2000 mL
250 mL
500 mL
5000 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09-841-305 5000 mL
09-841-300 100 mL
09-841-301 250 mL
09-841-302 500 mL
09-841-303 1000 mL
09-841-304 2000 mL
09-841-306 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 09-841-305 Supplier DWK Life Sciences Supplier No. 218017312
Add to Cart
Add to Cart

DURAN™ Original GL 45 Laboratory Bottles complete with High Temperature Closures.

DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, clear, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN Original GL 45 laboratory bottles are available complete with high temperature screw caps and pouring rings. The red polybutylene terephthalate (PBT) GL 45 threaded caps with PTFE faced silicone cap liner and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. All components are fully autoclavable at 121 or 134°C and suitable for dry heat sterilization up to 180°C. All components are fully compliant with current international food contact regulations such as (EU) No 10/2011.

  • Supplied complete with red polybutylene terephthalate (PBT) GL 45 threaded cap with PTFE faced silicone liner and ETFE pouring ring for drip free pouring.
  • Supplied complete with high temperature PBT screw caps and ETFE pouring rings.
  • The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel.
  • Permanent Retrace Code on each bottle for full manufacturing lot traceability.
  • Suitable for long-term storage.
  • All components are fully autoclavable at 121 or 134°C.
  • All components are suitable for dry heat sterilization up to 180°C.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5000 mL
For Use With (Application) Suitable for dry heat sterilization; Autoclaving of liquid media; Storage of corrosive reagents; Sampling; Storage of media and reagents; Transport and shipping
Includes With high temperature resistant PBT screw cap and ETFE pouring ring
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Thread Size (Metric) GL 45
Closure Material PBT with ETFE Pouring Ring
Diameter (Metric) 182 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.