Learn More
DWK LIFE SCIENCES™ DURAN™ Original Laboratory Bottle, Amber, with DIN 168-1 Thread, USP / EP (3.2.1) Light Transmission, Graduated, Bottle Only
Shop All DWK Life Sciences Products

Description
- Moulded from high-purity, amber USP/EP Type I DURAN™ borosilicate glass 3.3 for excellent thermal performance and high chemical resistance
- Fused external amber layer provides UV protection up to approx. 500 nm, meeting USP/EP (3.2.1) light transmission standards for the lifetime of the bottle
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring
- Easy-to-read graduated volume scale and marking field in fired-on, highly durable white ceramic enamel
- Permanent Retrace Code on each bottle for manufacturing lot traceability
- Complies with ISO 4796-1 design standards for a long working life
- Sizes 10 and 25 mL feature a specially shaped glass edge for improved pouring without requiring an additional plastic pouring ring
- All components fully autoclavable at 121°C or 134°C
Specifications
Specifications
| Capacity (Metric) | 20 L |
| Color | Translucent Amber |
| For Use With (Application) | Preparation of light-sensitive materials; Storage of light-sensitive materials; Transport and shipping of light-sensitive materials; Autoclaving of media and reagents; Culturing vessel |
| Material | Borosilicate Glass (3.3) |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
| Thread Size (Metric) | GL 45 |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Threaded |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.