Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated
SDP

Catalog No. 09841201 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
15 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09-841-201 15 L
09-841-184 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-841-201 Supplier DWK LIFE SCIENCES™ Supplier No. 218018855
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated. DURAN™ Original Laboratory Bottle is the gold standard for multifunctional glass laboratory bottles. Moulded from USP/EP Type I borosilicate glass 3.3, it offers outstanding chemical and thermal resistance, graduated scale

  • Moulded from high-purity, clear USP/EP Type I borosilicate glass 3.3 for excellent thermal performance and high chemical resistance
  • Complies with ISO 4796-1 design standard for a long working life
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring
  • Easy-to-read graduated volume scale and marking field in fired-on, highly durable permanent white enamel
  • Permanent Retrace Code on each bottle with certificate for manufacturing lot traceability
  • Supplied complete with iconic blue polypropylene cap and drip-free pouring ring rated for use up to 140°C
  • Fully autoclavable at 121°C or 134°C - all components including cap and pouring ring
  • Suitable for long-term storage of media, chemicals, and solvents
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 15 L
Color Clear
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Includes Blue Polypropylene Screw Cap and Pouring Ring
Material Borosilicate Glass (3.3)
No. per Pack 1 Ea.
Quantity 1 Ea.
Sterility Non-sterile
Thread Size (Metric) GL 45
Closure Color Blue
Closure Included Yes
Graduated Yes
Mouth Threaded
Fit for Purpose Certified Quality
Height (Metric) 450 mm
Closure Liner Linerless
Closure Material Polypropylene
Closure Size GL 45
Diameter (Metric) 268 mm
Autoclavable Autoclavable
Capacity (English) 3.96 gal.
Shape Round
Disposable Reusable
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.