Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated, Bottle Only
SDP

Catalog No. 09841163 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
150 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
09-841-163 150 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-841-163 Supplier DWK LIFE SCIENCES™ Supplier No. 218012906
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated, Bottle Only - the gold standard since 1972. USP/EP Type I borosilicate glass 3.3, standardised GL thread, autoclavable at 121 or 134°C, with graduated scale and lot traceability.

  • Moulded from high-purity, clear USP/EP Type I borosilicate glass 3.3 for excellent thermal performance and high chemical resistance
  • Conforms to ISO 4796-1 design for a long working life
  • Standardised GL thread and matching blue polypropylene cap for particularly tight sealing and simple, clean pouring
  • Drip-free polypropylene pouring ring included; suitable for use up to 140°C
  • Easy-to-read graduated volume scale and marking field in fired-on, highly durable white enamel print
  • Permanent Retrace Code on each bottle with certificate for manufacturing lot traceability
  • Fully autoclavable at 121°C or 134°C
  • Suitable for long-term storage of culture media, chemicals, and solvents
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 150 mL
Color Clear
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 45
Closure Included No
Graduated Yes
Mouth Threaded
Closure Size GL 45
Diameter (Metric) 62 mm
Autoclavable Autoclavable
Capacity (English) 5 oz.
Shape Round
Disposable Reusable
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.