Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated
SDP

Catalog No. 02900127 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-900-127 250 mL
02-900-128 500 mL
02-900-126 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-900-127 Supplier DWK Life Sciences Supplier No. 1127076
Add to Cart
Add to Cart

Glass laboratory bottles with superior thermal and chemical resistance.

The DURAN™ premium GL 45 laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. The premium bottles are ideal for high temperature applications: the thermal shock resistance of the glass bottle is certified for a 160°C differential, and is combined with a high temperature fluoropolymer (PFA) screw cap and pouring ring that are suitable for use up to 200°C. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature a double graduated scale for filling volume and a reversed scale for pouring. The marking field and Retrace Code for manufacturing lot traceability are printed in permanent white enamel.

  • Additional graduation marks and an opposite scaling (accurate to ± 5 %) simplify the reading of the filling level as well as the quantity withdrawn
  • Colourant free product
  • Reliable sealing due to PTFE-coated cap liner
  • TÜV tested thermal shock resistance of 160 K
  • Complete with PFA pouring ring and cap
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage and transport of media and reagents; Shipping
Includes Premium Screw Cap and Pouring Ring
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Thread Size (Metric) GL 45
Closure Material PFA/PFA, PTFE/Silicone Layer
Diameter (Metric) 70 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.