Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated
SDP

Catalog No. 02900128 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-900-128 500 mL
02-900-127 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-900-128 Supplier DWK LIFE SCIENCES™ Supplier No. 1127077
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated is moulded from USP/EP Type I borosilicate glass 3.3 with TÜV-certified 160 K thermal shock resistance and a PFA cap rated to 200°C.

  • Moulded from high-purity, clear USP/EP Type I borosilicate glass 3.3 with excellent thermal performance and high chemical resistance
  • TÜV-tested thermal shock resistance certified for a 160°C (160 K) differential
  • Complete with high-temperature PFA (fluoropolymer) screw cap and pouring ring rated for use up to 200°C
  • Standardised GL 45 thread and matching cap system for particularly tight sealing and simple, clean pouring
  • Double graduated scale for filling volume and a reversed scale for pouring (accurate to ± 5%)
  • Marking field and Retrace Code for manufacturing lot traceability printed in permanent white enamel
  • Colourant-free product; PTFE-coated cap liner ensures reliable sealing
  • Robust ISO 4796-1 design for a long working life
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Clear
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage and transport of media and reagents; Shipping
Includes Premium Screw Cap and Pouring Ring
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 45
Closure Included Yes
Graduated Yes
Mouth Narrow
Closure Material PFA/PFA, PTFE/Silicone Layer
Closure Size GL 45
Diameter (Metric) 86 mm
Autoclavable Autoclavable
Capacity (English) 17 oz.
Shape Square
Disposable Reusable
Product Type Laboratory Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.