Learn More
DWK Life Sciences DURAN™ Premium Screw Cap Replacement Cap Liner
SDP
Catalog No. 02900134 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 25
Catalog No. Thread Size (Metric)
02-900-134 GL 25
1 options
Catalog No. 02-900-134 Supplier DWK Life Sciences Supplier No. 292481407
Add to Cart
Add to Cart
The DURAN™ Premium Screw Cap Replacement Cap Liner provides a reliable, tight seal with high temperature and chemical resistance, making it ideal for pharmaceutical and demanding laboratory processes including sterilisation and depyrogenation.
  • High purity fluoropolymer (PFA) construction without any colourants
  • USP Class VI pharmaceutical grade materials meeting FDA, USP, EP and JP requirements
  • High temperature resistance suitable for hot-air/dry-heat sterilisation and depyrogenation at up to 200°C
  • Excellent chemical resistance for use in chemically demanding processes
  • PTFE-coated silicone cap liner provides a reliable and tight seal for long-term storage and transport of sensitive products
  • Compatible with GL 25 and GL 45 thread sizes for versatile use across DURAN bottle ranges
  • Replacement PTFE-faced silicone cap liners (platinum-cured silicone) available separately
  • Matching PFA pouring ring for drip-free pouring available separately
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless
Description Replacement cap liner
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Reservoir cap for supplying laboratory instruments; Port assemblies for spinner flasks, and cell culture vessels; Single port vent for autoclaving media in large bottles; Transport and shipping
Material PFA, PTFE, Silicone
Quantity 10/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 25
Diameter (Metric) 23.5 mm
Autoclavable Autoclavable
Disposable Reusable
Product Type Cap Sealing Disc