Learn More
DWK Life Sciences DURAN™ PURE Premium Cap, with Lip Seal
SDP
Catalog No. 02913032 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 45
Catalog No. Thread Size (Metric)
02-913-032 GL 45
1 options
Catalog No. 02-913-032 Supplier DWK Life Sciences Supplier No. 898862802
Add to Cart
Add to Cart
High-quality closure developed for the needs of the pharmaceutical and biotech industry
High-quality plastic screw cap with GL 45 thread size, developed for the needs of the pharmaceutical industry. The DURAN™ PURE Premium Lip Seal Cap is manufactured in clean rooms and made from PFA with lip seal design. Designed for use with DURAN™ PURE GL 45 Bottles. The screw cap is compatible with autoclaving and other common sterilization methods. DWK Life Sciences provides CoAs with each delivery, assures 100% traceability and change control management. The DURAN™ PURE Premium Lip Seal Cap is available with the documentation required by regulatory authorities and meets the requirements of the international pharmacopoeias.
  • High cleanliness due to manufacturing in clean rooms
  • Single batch delivery
  • Certificate of Analysis (CoA)
  • Visual and dimensional inspection
  • Change control management and information of customer in case of changes
  • 100% traceability of all manufacturing steps
  • Extractables report available
  • Drug Master File
  • USP <88> Class VI Pharmaceutical grade materials
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
For Use With (Application) Manufacturing of pharmaceutical drugs and biotech products; Storage and transport of APIs, bulk drug intermediates, ingredients and media
Material PFA
Quantity 5/Pk.
Thread Size (Metric) GL 45
Neck Finish 98 mm
Diameter (Metric) 53 mm
Autoclavable Autoclavable