Learn More
DWK Life Sciences DURAN™ Replacement Cap Liner, PTFE-Coated, Peroxide cured silicone
SDP
Catalog No. 02900139 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 18
GL 25
GL 32
GL 45
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
02-900-139 GL 18
12-141-601 GL 45
02-900-140 GL 25
02-900-141 GL 32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-900-139 Supplier DWK Life Sciences Supplier No. 292481107
Add to Cart
Add to Cart
Cap liners with PTFE faced silicone.
Replacement cap liners suitable for PBT screw caps and tamper-evident screw caps. Heat resistance to 130°C (steam) and 200°C (dry heat). Excellent chemical resistance due to PTFE coating. Silicon is peroxide catalysed.
  • replacement, cap, liner, silicone, seal, sealing, vmq, ptfe, coated, screw, cap
  • Excellent chemical resistance due to PTFE coating
  • Cap liner forms a tight seal with bottle Peroxide catalysed silicone
  • Peroxide catalysed silicone
  • Suitable for PBT screw caps and tamper-evident screw caps
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Replacement cap liner for red PBT caps
Material Silicone, PTFE, VMQ
Quantity 10/Pk.
Thread Size (Metric) GL 18
Diameter (Metric) 16.8 mm
Autoclavable Autoclavable