Learn More
DWK Life Sciences DURAN™ Replacement Cap Liner, PTFE-Coated, Peroxide cured silicone
SDP
Catalog No. 02900140 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 18
GL 25
GL 32
GL 45
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
02-900-140 GL 25
12-141-601 GL 45
02-900-139 GL 18
02-900-141 GL 32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-900-140 Supplier DWK Life Sciences Supplier No. 292481304
Only null left
Add to Cart
Add to Cart
Cap liners with PTFE faced silicone.
Replacement cap liners suitable for PBT screw caps and tamper-evident screw caps. Heat resistance to 130°C (steam) and 200°C (dry heat). Excellent chemical resistance due to PTFE coating. Silicon is peroxide catalysed.
  • replacement, cap, liner, silicone, seal, sealing, vmq, ptfe, coated, screw, cap
  • Excellent chemical resistance due to PTFE coating
  • Cap liner forms a tight seal with bottle Peroxide catalysed silicone
  • Peroxide catalysed silicone
  • Suitable for PBT screw caps and tamper-evident screw caps
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Replacement cap liner for red PBT caps
Material Silicone, PTFE, VMQ
Quantity 10/Pk.
Thread Size (Metric) GL 25
Diameter (Metric) 23.5 mm
Autoclavable Autoclavable