Learn More
DWK Life Sciences DURAN™ Replacement Parts for HPLC Screw Cap GL 45

Shop All DWK Life Sciences Products


Description
- Replacement parts kit for the DURAN™ HPLC screw cap GL 45 four-port system
- Includes M8 screw caps and silicone seals compatible with 1.6 mm and 3.2 mm tubing, plus blind silicone seals for unused ports
- Silicone seals ensure a secure, leak-free connection at each port
- Unused ports can be sealed with included blind silicone seals to maintain system integrity
- Compatible with the DURAN™ HPLC bottle system designed for pressure and vacuum resistant closed-system liquid transfer
- Supports safe transfer of liquid media within a closed and sterile system, reducing evaporation
Specifications
Specifications
| Color | Blue |
| Description | Replacement parts for HPLC screw cap (includes M8 caps and 1.6, 3.2 mm and blind silicone seals) |
| For Use With (Application) | Pressure and vacuum resistant bottle for HPLC applications; Safe transfer of liquid media within a closed and sterile system; Supply of solvents to HPLC equipment |
| Material | Borosilicate Glass (3.3) |
| Quantity | 1 Ea. |
| Sterility | Non-sterile |
| Thread Size (Metric) | GL 45 |
| Closure Material | Polypropylene with Silicone Seal |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.