Learn More
DWK Life Sciences DURAN™ Screw Caps for GLS 80™ Wide Neck Glass Bottles
Shop All DWK Life Sciences Products

Description
- High leak tightness achieved through PTFE-coated silicone seal
- Greater chemical resistance compared to standard polypropylene (PP) screw caps
- Compatible with DURAN GLS 80 wide neck glass bottles
- Matching ETFE pouring ring available separately for clean, drip-free use
Specifications
Specifications
| Color | Red |
| Material | PBT |
| Sterility | Non-sterile |
| Thread Size (Metric) | GL 14 |
| For Use With (Equipment) | GLS 80 wide neck glass bottles |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Product Type | Screw Cap |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.