Learn More
DWK Life Sciences DURAN™ Silicone Septum Seal, Peroxide cured silicone, Septum for Piercing, Use with Open Topped Screw Cap
Shop All DWK Life Sciences Products

Description
Suitable for PBT open topped screw cap with central aperture Heat resistance: 130°C (steam) and 200°C (dry heat). Silicon is peroxide catalysed.
- Compatible with open topped GL screw caps with central aperture
- Compatible with GL bottles
- Peroxide catalysed silicone
Specifications
Specifications
| For Use With (Application) | Liquid transfer from HPLC solvent bottles; Aseptic reagent delivery or culture sampling using a syringe and needle; Reservoir cap for supplying laboratory instruments |
| Material | Silicone |
| Quantity | 100/Pk. |
| Thickness (Metric) | 2 mm |
| Thread Size (Metric) | GL14 |
| Diameter (Metric) | 12 mm |
| Autoclavable | Autoclavable |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.