Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences DURAN™ Silicone Septum Seal, Peroxide Cured Silicone, Septum for Piercing, Use with Open Topped Screw Cap
SDP

Catalog No. 02900143 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL14
GL 32
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
02-900-143 GL 32
02-900-142 GL14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-900-143 Supplier DWK Life Sciences Supplier No. 292460803
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The DURAN™ Silicone Septum Seal, Peroxide Cured Silicone, Septum for Piercing, Use with Open Topped Screw Cap is suitable for PBT open topped screw caps with central aperture. Peroxide catalysed silicone offers heat resistance to 130°C (steam) and 200°C (dry heat).

  • Compatible with open topped GL screw caps with central aperture
  • Compatible with DURAN GL bottles
  • Peroxide catalysed silicone construction
  • Heat resistant to 130°C (steam) and 200°C (dry heat)
  • Designed for injection or removal of media via piercing
TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Application) Liquid transfer from HPLC solvent bottles; Aseptic reagent delivery or culture sampling using a syringe and needle; Reservoir cap for supplying laboratory instruments
Material Silicone
Quantity 100/Pk.
Sterility Non-sterile
Thickness (Metric) 2 mm
Thread Size (Metric) GL 32
Diameter (Metric) 29 mm
Autoclavable Autoclavable
Shape Round
Product Type Septa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.