Learn More
DWK Life Sciences DURAN™ TILT GL 56 Screw Cap Membrane Vented
SDP
Catalog No. 09841310 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 56
Catalog No. Thread Size (Metric)
09-841-310 GL 56
1 options
Catalog No. 09-841-310 Supplier DWK Life Sciences Supplier No. 291185609
Add to Cart
Add to Cart
The DURAN™ TILT GL 56 Screw Cap Membrane Vented enables safe pressure equalisation during autoclaving via a 0.2‐μm ePTFE membrane, maintaining sterility while allowing bidirectional sterile gas exchange for secure media storage.
  • Designed for use with DURAN™ TILT laboratory bottles for sterilising and storing media or buffer solutions
  • 0.2‐μm ePTFE membrane permits automatic sterile gas exchange in two directions, allowing safe pressure equalisation during the autoclave cycle
  • Microporous membrane structure blocks penetration of liquids and solids, keeping bottle contents sterile
  • Increased gas exchange rate due to thin ePTFE membrane combined with larger ventilation openings
  • ePTFE membrane and polypropylene mesh support are ultrasonically welded, forming a durable seal
  • ePTFE membrane conforms to USP 121°C Class VI; supported by polypropylene mesh
  • Cap can be tightly closed during autoclaving, no need to leave cap loose during the sterilisation cycle
  • Cost-effective reusable design; replacement recommended after 3 to 5 autoclave cycles or if membrane becomes damaged or contaminated
  • GL 56 thread size
TRUSTED_SUSTAINABILITY
Specifications
Color White
For Use With (Application) Pressure equalisation during steam sterilisation/autoclaving of media or buffers; Sterile gas exchange for aerobic microbial, shaker and suspension cell cultures; Autoclaving of media and buffers; Short-term storage of sterilised media; Storage of gas generating materials
Material Polypropylene, PTFE
Quantity 4/Pk.
Sterility Non-sterile
Thread Size (Metric) GL 56
Neck Finish 91 mm
Diameter (Metric) 62 mm
Autoclavable Autoclavable
Disposable Reusable
Product Type Bottle Cap