Description
- DURAN™ borosilicate glass construction for excellent chemical and thermal resistance
- Fused rim design provides a smooth, reinforced edge for enhanced durability and safe handling
- Suitable for use as a beaker cover, evaporation dish, or general laboratory vessel
- Transparent glass allows easy visual monitoring of contents
- Compatible with standard laboratory cleaning and sterilisation procedures
Specifications
Specifications
| Bottom Shape | Concave |
| Material | Borosilicate Glass (3.3) |
| Sterility | Non-sterile |
| Type | Watch Glass |
| Edges | Fused rim |
| Diameter (Metric) | 250 mm |