Learn More
DWK Life Sciences DURAN™ YOUTILITY™ Label, Printable Self-Adhesive
Shop All DWK Life Sciences Products

Description
- High temperature performance range makes them ideal for use in fridges, freezers, autoclaves, incubators, and water baths
- Can be printed with office printers, copiers, or marked with technical lab pens
- High adhesive strength; labels are peelable and easy to remove, saving time during cleaning
- Free easy-to-use Label Creator web app available for individual label design
- Chemically resistant to many typical laboratory chemicals and solvents
- Non-hazardous and free of toxic heavy metals
Specifications
Specifications
| Autoclavable | Autoclavable |
| Adhesive Type | Self-adhesive |
| Coating | Polyester |
| Color | White |
| Format | Label |
| Material | Polyester |
| Product Type | Label |
| Sterility | Non-sterile |
| For Use With (Application) | Labelling on YOUTILITY bottles; Sample labelling |
| Quantity | 1 x 100 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.