Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK LIFE SCIENCES™ DURAN™ YOUTILITY™ Screw Cap GL 45
SDP

Catalog No. 09841069 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
GL 45
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
09-841-069 GL 45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-841-069 Supplier DWK LIFE SCIENCES™ Supplier No. 292292802
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The DURAN™ YOUTILITY™ Screw Cap GL 45 features an optimised sealing system for a liquid-tight seal, ergonomic grooves for easy tightening, a faster-opening thread compatible with DIN GL 45, integrated retrace code for full traceability, and a dedicated labelling area.

  • Ergonomically shaped screw cap with optimised grooves and ridges for efficient and easier tightening or removal
  • Faster opening and closing thread is fully compatible with DIN GL 45 bottle threads
  • Screw cap has an integrated retrace code to provide full product traceability
  • Pre-defined labelling area on the cap is compatible with the dedicated DURAN™ self-adhesive YOUTILITY labels
  • Optimised cap sealing system ensures a liquid-tight seal
TRUSTED_SUSTAINABILITY

Specifications

Color Blue-Green
Includes Pre-defined labelling area; Screw cap has an integrated retrace code
Material Polypropylene
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Threaded
For Use With (Equipment) DIN GL 45 bottle threads
Closure Size GL 45
Autoclavable Autoclavable
Shape Straight-sided
Disposable Reusable
Product Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.