Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR double walled jacke1EA
Catalog No. 502908893
Encompass
The DURAN double walled wide mouth bottles GLSA80 incorporate an integral jacket that isolates the contents from the external environment. Heated or cooled liquids can be circulated through the jacket to control the temperature within the screw topped DURAN bottle. The DURAN double walled bottles offer a sealable and more flexible alternative to open topped jacketed beakers.
Catalog No. 50-290-8893 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7423881EA
The DURAN double walled wide mouth bottles GLSA80 incorporate an integral jacket that isolates the contents from the external environment. Heated or cooled liquids can be circulated through the jacket to control the temperature within the screw topped DURAN bottle. The DURAN double walled bottles offer a sealable and more flexible alternative to open topped jacketed beakers.