Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR GL 45 connection sy
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z6803972EA
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available 1.6 mm 3.0 mm 3.2 mm and 6.0 mm that can be connected.Sterile pressure equalization is possible through use of membrane filter.