Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR GLS 80 wide mouth g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z67411710EA
Encompass
The wide opening for safe and easy handling of powders and liquids makes the Duran GLS 80 bottle a versatile partner for all routine laboratory applications.Duran glass elliminates ion leaching problemsSturdy design enhances safety and service lifeThermal properties allow sterilisation by autoclaveConvenient and easy to read graduationsSupplied with blue quick-release cap and pouring ring that is autoclavable up to 140C
Catalog No. 50-290-5663