Learn More
Sigma Aldrich Fine Chemicals Biosciences DuranR GLS 80 wide mouth g1EA
Catalog No. 502906002
Encompass
The wide opening for safe and easy handling of powders and liquids makes the Duran GLS 80 bottle a versatile partner for all routine laboratory applications.Duran glass elliminates ion leaching problemsSturdy design enhances safety and service lifeThermal properties allow sterilisation by autoclaveConvenient and easy to read graduationsSupplied with blue quick-release cap and pouring ring that is autoclavable up to 140C
Catalog No. 50-290-6002 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6741331EA
The wide opening for safe and easy handling of powders and liquids makes the Duran GLS 80 bottle a versatile partner for all routine laboratory applications.Duran glass elliminates ion leaching problemsSturdy design enhances safety and service lifeThermal properties allow sterilisation by autoclaveConvenient and easy to read graduationsSupplied with blue quick-release cap and pouring ring that is autoclavable up to 140C